Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270483.1 | NP_001257412.1 | 3 | 869 | ||
NM_003312.5 | NP_003303.2 | 3 | 901 | |||
GenBank mRNA | AK000862.1 | - | 3 | 895 | ||
AK091874.1 | - | 2 | 1521 | |||
AK223521.1 | - | 3 | 895 | |||
AK301300.1 | - | 4 | 736 | |||
BC010148.2 | - | 3 | 878 | 4329727 | ||
CR456598.1 | - | 2 | 904 | |||
CR456998.1 | - | 2 | 826 | |||
D87292.1 | - | 3 | 874 |