Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144766.2 | NP_001138238.1 | 4, 5 | 450 | ||
NM_003315.3 | NP_003306.3 | 4, 5 | 682 | |||
NR_029431.1 | 3, 4 | 593 | ||||
XM_011525167.2 | XP_011523469.1 | 5, 6 | 612 | |||
XM_011525168.2 | XP_011523470.1 | 5, 6 | 572 | |||
XM_011525169.2 | XP_011523471.1 | 4, 5 | 424 | |||
XM_017024994.1 | XP_016880483.1 | 5, 6 | 606 | |||
GenBank mRNA | AB209834.1 | - | 4, 5 | 368 | ||
AK291047.1 | - | 4, 5 | 383 | |||
AK298860.1 | - | 4, 5 | 398 | |||
AK308016.1 | - | 4, 5 | 436 | |||
AK316081.1 | - | 3, 4 | 319 | |||
BC003601.1 | - | 3, 4 | 305 | 3543074 | ||
BC011837.2 | - | 4, 5 | 401 | 3945721 | ||
BC033772.1 | - | 4, 5 | 427 | 3924081 | ||
BP378441.1 | - | 4, 5 | 431 | |||
BX647209.1 | - | 4, 5 | 638 | |||
U46571.1 | - | 4, 5 | 397 |