Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080850.2 | NP_001074319.1 | 4 | 384 | ||
XM_011542067.2 | XP_011540369.1 | 5 | 1316 | |||
XM_011542068.2 | XP_011540370.1 | 5 | 1321 | |||
XM_011542069.2 | XP_011540371.1 | 4 | 1175 | |||
XM_011542070.2 | XP_011540372.1 | 3 | 342 | |||
XM_017002225.1 | XP_016857714.1 | 4 | 401 | |||
XM_017002226.1 | XP_016857715.1 | 5 | 712 | |||
GenBank mRNA | AK302075.1 | - | 6 | 568 | ||
AL832217.1 | - | 10 | 1279 |