Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110781.2 | NP_001104251.1 | 9 | 1685 | ||
NM_001290264.1 | NP_001277193.1 | 10 | 2103 | |||
XM_011542072.1 | XP_011540374.1 | 5 | 1046 | |||
GenBank mRNA | AB007916.3 | - | 9 | 1623 | ||
BC110653.1 | - | 9 | 1661 | 4392812 | ||
CR936618.1 | - | 10 | 1823 |