Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544184.2 | XP_011542486.1 | 1 | 207 | ||
GenBank mRNA | AK090902.1 | - | 1 | 207 |