Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM200C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005258130.4 | XP_005258187.1 | 1 | 524 | ||
XM_011525729.2 | XP_011524031.1 | 1 | 524 |