Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099218.2 | NP_001092688.1 | 1 | 312 | ||
NM_001321233.1 | NP_001308162.1 | 5 | 670 | |||
XM_011533084.1 | XP_011531386.1 | 1 | 312 | |||
GenBank mRNA | AK310498.1 | - | 6 | 917 | ||
DQ860102.1 | - | 1 | 312 |