Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP24-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102705.1 | 2 | 855 | |||
NR_102706.1 | 2 | 818 | ||||
GenBank mRNA | BQ953249.1 | - | 2 | 471 | ||
BU159548.1 | - | 1 | 98 |