Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244938.1 | NP_001231867.1 | 3 | 357 | ||
NM_003329.3 | NP_003320.2 | 4 | 417 | |||
GenBank mRNA | AF065241.1 | - | 3 | 149 | ||
AF085844.1 | - | 4 | 224 | |||
AF313911.1 | - | 4 | 222 | |||
AK289508.1 | - | 4 | 338 | |||
AW160756.1 | - | 4 | 417 | |||
AY004872.1 | - | 4 | 222 | |||
BC003377.1 | - | 4 | 269 | 2901084 | ||
BC054866.1 | - | 4 | 288 | 6463280 | ||
BQ774337.1 | - | 2 | 466 | |||
BX647247.1 | - | 3 | 258 | |||
CR407665.1 | - | 4 | 212 | |||
JQ313905.1 | - | 4 | 212 | |||
X77584.1 | - | 4 | 292 |