Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101417.3 | NP_001094887.1 | 6 | 832 | ||
NM_001101426.3 | NP_001094896.1 | 7 | 982 | |||
XM_006715770.3 | XP_006715833.1 | 7 | 1152 | |||
XM_011515499.2 | XP_011513801.1 | 7 | 1127 | |||
XM_011515500.2 | XP_011513802.1 | 6 | 1022 | |||
XM_011515508.2 | XP_011513810.1 | 7 | 1484 | |||
XM_011515509.2 | XP_011513811.1 | 7 | 2333 | |||
XM_017012575.1 | XP_016868064.1 | 7 | 1127 | |||
XM_017012577.1 | XP_016868066.1 | 6 | 1244 | |||
XM_017012578.1 | XP_016868067.1 | 5 | 1659 | |||
GenBank mRNA | DY654823.1 | - | 4 | 235 |