Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286389.1 | NP_001273318.1 | 4 | 2468 | ||
XM_011536745.2 | XP_011535047.1 | 5 | 3195 | |||
XM_011536750.2 | XP_011535052.1 | 5 | 2380 | |||
GenBank mRNA | AF462446.1 | - | 1 | 778 | ||
BC047917.1 | - | 1 | 425 |