Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003334.3 | NP_003325.2 | 9 | 1113 | ||
NM_153280.2 | NP_695012.1 | 9 | 1041 | |||
XM_005272649.1 | XP_005272706.1 | 10 | 1491 | |||
XM_011543954.2 | XP_011542256.1 | 10 | 1133 | |||
XM_017029777.1 | XP_016885266.1 | 9 | 2208 | |||
XM_017029778.1 | XP_016885267.1 | 10 | 3382 | |||
XM_017029779.1 | XP_016885268.1 | 9 | 3890 | |||
XM_017029780.1 | XP_016885269.1 | 9 | 998 | |||
XM_017029781.1 | XP_016885270.1 | 9 | 1046 | |||
GenBank mRNA | BC013041.2 | - | 9 | 980 | 3543021 | |
BC020261.1 | - | 6 | 675 | |||
M58028.1 | - | 9 | 1019 | |||
X52897.1 | - | 2 | 185 | |||
X56976.1 | - | 9 | 922 |