Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003358.2 | NP_003349.1 | 3 | 776 | ||
XM_005252186.4 | XP_005252243.1 | 4 | 799 | |||
XM_017015107.1 | XP_016870596.1 | 3 | 435 | |||
GenBank mRNA | AK314847.1 | - | 3 | 381 | ||
BC038711.1 | - | 3 | 777 | 5295561 | ||
D50840.1 | - | 3 | 616 |