Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003380.3 | NP_003371.2 | 9, 10 | 1766 | ||
XM_006717500.1 | XP_006717563.1 | 8, 9 | 2841 | |||
GenBank mRNA | AF328728.1 | - | 8, 9 | 1482 | ||
AK056766.1 | - | 9, 10 | 1839 | |||
AK091813.1 | - | 9, 10 | 1457 | |||
AK093924.1 | - | 5, 6 | 2297 | |||
AK097336.1 | - | 9, 10 | 1462 | |||
AK098444.1 | - | 9, 10 | 1336 | |||
AK222482.1 | - | 8, 9 | 1496 | |||
AK222507.1 | - | 8, 9 | 1498 | |||
AK222602.1 | - | 8, 9 | 1498 | |||
AK290643.1 | - | 8, 9 | 1378 | |||
BC000163.2 | - | 8, 9 | 1477 | 2985712 | ||
BC030573.2 | - | 8, 9 | 1477 | 3637336 | ||
BC066956.1 | - | 9, 10 | 1554 | 4823475 | ||
CR407690.1 | - | 8, 9 | 1353 | |||
EU794672.1 | - | 8, 9 | 1498 | |||
KU178388.1 | - | 8, 9 | 1353 | |||
KU178389.1 | - | 9, 10 | 970 | |||
M25246.1 | - | 8, 9 | 857 | |||
X56134.1 | - | 8, 9 | 1396 | |||
Z19554.2 | - | 8, 9 | 1475 |