Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPREB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303509.1 | NP_001290438.1 | 2 | 156 | ||
NM_007128.3 | NP_009059.1 | 2 | 170 | |||
GenBank mRNA | BC017922.1 | - | 2 | 162 | ||
BG502270.1 | - | 2 | 156 | |||
CR456609.1 | - | 2 | 147 |