Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018727.5 | NP_061197.4 | 4 | 719 | ||
NM_080704.3 | NP_542435.2 | 5 | 836 | |||
NM_080705.3 | NP_542436.2 | 4 | 762 | |||
NM_080706.3 | NP_542437.2 | 3 | 1088 | |||
GenBank mRNA | AF196175.1 | - | 4 | 774 | ||
AF196176.1 | - | 4 | 762 | |||
AJ272063.2 | - | 4 | 719 | |||
AJ277028.1 | - | 3 | 1088 | |||
AL136801.1 | - | 5 | 832 | |||
AY131289.1 | - | 3 | 561 | |||
AY986821.1 | - | 3 | 561 | |||
BC132820.1 | - | 3 | 1040 | 40146842 | ||
BC136633.1 | - | 3 | 1040 | 9020623 | ||
DQ177332.1 | - | 2 | 648 | |||
DQ177333.1 | - | 3 | 578 | |||
DQ898279.1 | - | 3 | 561 |