Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004184.3 | NP_004175.2 | 11 | 1672 | ||
NM_173701.1 | NP_776049.1 | 11 | 1448 | |||
NM_213645.1 | NP_998810.1 | 10 | 1276 | |||
NM_213646.1 | NP_998811.1 | 10 | 1500 | |||
XM_005268044.3 | XP_005268101.1 | 11 | 1400 | |||
XM_006720249.3 | XP_006720312.1 | 12 | 1634 | |||
XM_011537133.2 | XP_011535435.1 | 13 | 1646 | |||
XM_011537135.2 | XP_011535437.1 | 12 | 1530 | |||
XM_011537136.2 | XP_011535438.1 | 11 | 1366 | |||
XM_017021627.1 | XP_016877116.1 | 12 | 1538 | |||
XM_017021628.1 | XP_016877117.1 | 12 | 1669 | |||
XM_017021629.1 | XP_016877118.1 | 13 | 1644 | |||
GenBank mRNA | AK025487.1 | - | 9 | 1076 | ||
AK056100.1 | - | 11 | 1910 | |||
AK124668.1 | - | 1 | 972 | |||
AK130698.1 | - | 7 | 779 | |||
AK291141.1 | - | 10 | 1252 | |||
BC017489.1 | - | 11 | 1548 | 3542671 | ||
BC095453.1 | - | 12 | 1559 | 30418785 | ||
BX248006.1 | - | 11 | 1392 | |||
M61715.1 | - | 11 | 1478 | |||
M77804.1 | - | 10 | 1333 | |||
X59892.1 | - | 11 | 1402 | |||
X62570.1 | - | 10 | 1353 |