Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014206.3 | NP_055021.1 | 3 | 280 | ||
GenBank mRNA | AF070661.1 | - | 3 | 280 | ||
AF085986.1 | - | 3 | 253 | |||
AF086763.1 | - | 3 | 276 | |||
AK291683.1 | - | 3 | 277 | |||
BC002750.2 | - | 3 | 251 | 3633279 | ||
BC015968.1 | - | 3 | 281 | 4073907 |