Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006522.3 | NP_006513.1 | 2 | 503 | ||
GenBank mRNA | AB059570.1 | - | 2 | 293 | ||
AK075522.1 | - | 2 | 387 | |||
AY009401.1 | - | 2 | 511 | |||
BC004329.1 | - | 2 | 291 | 3630498 | ||
BT007456.1 | - | 2 | 288 |