Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048212.3 | NP_001041677.1 | 1 | 525 | ||
GenBank mRNA | AY584596.1 | - | 1 | 525 | ||
BC146925.1 | - | 1 | 475 | 9056853 | ||
BC146926.1 | - | 1 | 475 | 9056854 |
Gene Symbol: AMDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145815.1 | NP_001139287.1 | 11 | 2003 | ||
XM_017023262.1 | XP_016878751.1 | 11 | 2655 | |||
XM_017023263.1 | XP_016878752.1 | 9 | 2643 | |||
XM_017023264.1 | XP_016878753.1 | 10 | 2586 | |||
XM_017023265.1 | XP_016878754.1 | 10 | 2553 | |||
XM_017023266.1 | XP_016878755.1 | 10 | 2601 | |||
XM_017023267.1 | XP_016878756.1 | 9 | 2597 | |||
GenBank mRNA | AK296877.1 | - | 11 | 1975 |