Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004630.3 | NP_004621.2 | 1, 2 | 482 | ||
NM_201995.2 | NP_973724.1 | 1, 2 | 482 | |||
NM_201997.2 | NP_973726.2 | 1, 2 | 482 | |||
NM_201998.2 | NP_973727.1 | 1, 2 | 482 | |||
NR_033649.1 | 1, 2 | 482 | ||||
NR_033650.1 | 1, 2 | 482 | ||||
XM_017018247.1 | XP_016873736.1 | 1, 2 | 197 | |||
XM_017018248.1 | XP_016873737.1 | 1, 2 | 197 | |||
XM_017018249.1 | XP_016873738.1 | 1, 2 | 681 | |||
XM_017018250.1 | XP_016873739.1 | 1, 2 | 407 | |||
GenBank mRNA | AK296237.1 | - | 1, 2 | 180 | ||
AK299705.1 | - | 1, 2 | 180 | |||
AK301803.1 | - | 1, 2 | 407 | |||
BC008080.2 | - | 1, 2 | 178 | 2961666 | ||
BC008724.2 | - | 1, 2 | 178 | 2961666 | ||
BC020217.1 | - | 1, 2 | 381 | 4869292 | ||
BC034451.1 | - | 1, 2 | 407 | |||
D26120.1 | - | 1, 2 | 412 | |||
D26121.1 | - | 1, 2 | 412 | |||
DC368015.1 | - | 1, 2 | 438 | |||
DC370979.1 | - | 1, 2 | 482 | |||
L49345.1 | - | 1, 2 | 368 | |||
L49380.1 | - | 1, 2 | 368 | |||
Y08765.1 | - | 1, 2 | 72 | |||
Y08766.1 | - | 1, 2 | 324 |