Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004339.3 | NP_004330.1 | 3 | 489 | ||
GenBank mRNA | AF149785.1 | - | 3 | 427 | ||
AK094882.1 | - | 3 | 222 | |||
AK290139.1 | - | 3 | 242 | |||
AK315626.1 | - | 3 | 304 | |||
BC000415.2 | - | 3 | 246 | 2961420 | ||
BC012858.1 | - | 3 | 272 | 3857505 | ||
BC019295.1 | - | 3 | 236 | 2823814 | ||
BC020983.1 | - | 3 | 253 | 3851447 | ||
BC031097.1 | - | 3 | 267 | 5288388 | ||
BC034250.1 | - | 3 | 261 | 3862830 | ||
BG721942.1 | - | 3 | 428 | |||
DC396020.1 | - | 3 | 490 | |||
Z50022.1 | - | 3 | 295 |