Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005181.3 | NP_005172.1 | 5 | 544 | ||
GenBank mRNA | AK096880.1 | - | 5 | 544 | ||
AK300670.1 | - | 5 | 438 | |||
AK300677.1 | - | 4 | 465 | |||
AK313254.1 | - | 5 | 513 | |||
BC004897.2 | - | 5 | 499 | 3532715 | ||
DA421884.1 | - | 5 | 544 | |||
EU794596.1 | - | 5 | 1144 |