Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300951.1 | NP_001287880.1 | 3 | 735 | ||
NM_003430.3 | NP_003421.2 | 4 | 831 | |||
XM_006722880.3 | XP_006722943.1 | 2 | 556 | |||
XM_011528265.2 | XP_011526567.1 | 4 | 792 | |||
XM_011528266.2 | XP_011526568.1 | 4 | 760 | |||
XM_017027235.1 | XP_016882724.1 | 4 | 785 | |||
XM_017027236.1 | XP_016882725.1 | 3 | 652 | |||
GenBank mRNA | AK291256.1 | - | 4 | 738 | ||
AK300294.1 | - | 3 | 621 | |||
L11672.1 | - | 4 | 719 |