Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF133 SIRNA Assays ›
Gene Symbol:
ZNF133
Gene Name:
zinc finger protein 133
Gene Aliases:
ZNF150, pHZ-13, pHZ-66
Chromosome Location:
Chr.20: 18288228 - 18316996 on Build GRCh38
UniGene ID:
Hs.472221
Species:
Human
siRNA ID
HSS187811
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF133
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083330.2
NP_001076799.1
4
2281
NM_001282995.1
NP_001269924.1
6
2517
NM_001282996.1
NP_001269925.1
6
2391
NM_001282997.1
NP_001269926.1
5
2442
NM_001282998.1
NP_001269927.1
4
2271
NM_001282999.1
NP_001269928.1
4
2284
NM_001283000.1
NP_001269929.1
5
2460
NM_001283001.1
NP_001269930.1
5
2447
NM_001283002.1
NP_001269931.1
5
2349
NM_001283003.1
NP_001269932.1
5
2422
NM_001283004.1
NP_001269933.1
5
2409
NM_001283005.1
NP_001269934.1
2
2045
NM_001283006.1
NP_001269935.1
3
2123
NM_001283007.1
NP_001269936.1
3
2141
NM_001283008.1
NP_001269937.1
4
2311
NM_003434.5
NP_003425.2
7
2693
XM_005260819.1
XP_005260876.1
4
2093
XM_005260820.4
XP_005260877.1
6
2350
XM_011529336.1
XP_011527638.1
7
2429
XM_011529337.1
XP_011527639.1
6
2301
XM_011529338.2
XP_011527640.1
6
2295
XM_011529339.1
XP_011527641.1
7
2429
XM_011529340.1
XP_011527642.1
2
1904
XM_017028041.1
XP_016883530.1
5
10194
XM_017028042.1
XP_016883531.1
5
3468
XM_017028043.1
XP_016883532.1
5
10191
XM_017028044.1
XP_016883533.1
7
10541
XM_017028045.1
XP_016883534.1
5
10609
XM_017028046.1
XP_016883535.1
5
10207
XM_017028047.1
XP_016883536.1
5
3561
XM_017028048.1
XP_016883537.1
4
3390
XM_017028049.1
XP_016883538.1
7
10551
XM_017028050.1
XP_016883539.1
7
2449
XM_017028051.1
XP_016883540.1
8
2625
XM_017028052.1
XP_016883541.1
6
2289
XM_017028053.1
XP_016883542.1
4
2323
XM_017028054.1
XP_016883543.1
5
2203
XM_017028055.1
XP_016883544.1
8
2619
XM_017028056.1
XP_016883545.1
10
2838
XM_017028057.1
XP_016883546.1
7
2406
XM_017028058.1
XP_016883547.1
8
2623
GenBank mRNA
AK123005.1
-
2
3859
AK291389.1
-
5
2199
AK294263.1
-
3
1872
AK295273.1
-
5
2121
AK295513.1
-
5
2185
AK313525.1
-
4
2020
AK316386.1
-
2
1799
BC001887.1
-
7
2499
3535533
BT007310.1
-
3
1940
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline