Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164530.1 | NP_001158002.1 | 4 | 318 | ||
NM_001289401.1 | NP_001276330.1 | 5 | 385 | |||
NM_001289402.1 | NP_001276331.1 | 5 | 748 | |||
NM_003436.3 | NP_003427.3 | 5 | 416 | |||
NM_007134.1 | NP_009065.1 | 4 | 354 | |||
XM_005259211.1 | XP_005259268.1 | 4 | 318 | |||
XM_006723362.3 | XP_006723425.1 | 5 | 424 | |||
XM_006723363.3 | XP_006723426.1 | 5 | 788 | |||
XM_017027240.1 | XP_016882729.1 | 4 | 741 | |||
XM_017027241.1 | XP_016882730.1 | 5 | 743 | |||
GenBank mRNA | AK094202.1 | - | 5 | 707 | ||
AK098011.1 | - | 4 | 318 | |||
AK295110.1 | - | 5 | 386 | |||
BC046434.1 | - | 5 | 380 | 4824328 | ||
BX647775.1 | - | 4 | 322 | |||
DB117846.1 | - | 4 | 318 |