Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001218.4 | NP_001209.1 | 5 | 898 | ||
NM_001293642.1 | NP_001280571.1 | 4 | 718 | |||
NM_206925.2 | NP_996808.1 | 5 | 898 | |||
NR_135511.1 | 5 | 1001 | ||||
GenBank mRNA | AF037335.1 | - | 5 | 623 | ||
AF051882.1 | - | 5 | 515 | |||
AK096845.1 | - | 4 | 454 | |||
AK315769.1 | - | 5 | 664 | |||
BC000278.1 | - | 5 | 620 | 3357439 | ||
BC011691.2 | - | 5 | 623 | 3344734 | ||
BC023981.1 | - | 5 | 638 | 4042589 | ||
BT006656.1 | - | 5 | 508 |