Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007148.4 | NP_009079.2 | 4 | 789 | ||
XM_006721571.3 | XP_006721634.1 | 4 | 4797 | |||
XM_006721572.3 | XP_006721635.1 | 4 | 4795 | |||
XM_006721573.3 | XP_006721636.1 | 4 | 530 | |||
GenBank mRNA | AB026054.1 | - | 4 | 643 | ||
AK226052.1 | - | 4 | 729 | |||
AK309372.1 | - | 4 | 789 | |||
AK313844.1 | - | 4 | 735 | |||
BC053989.1 | - | 4 | 704 | 5922474 |