Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF185 SIRNA Assays ›
Gene Symbol:
ZNF185
Gene Name:
zinc finger protein 185 (LIM domain)
Gene Aliases:
SCELL
Chromosome Location:
Chr.X: 152898132 - 152973481 on Build GRCh38
UniGene ID:
Hs.16622
Species:
Human
siRNA ID
HSS187836
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF185
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178106.1
NP_001171577.1
20
1888
NM_001178107.1
NP_001171578.1
19
1801
NM_001178108.1
NP_001171579.1
19
1795
NM_001178109.1
NP_001171580.1
18
1705
NM_001178110.1
NP_001171581.1
17
1615
NM_001178113.1
NP_001171584.1
11
1345
NM_001178115.1
NP_001171586.1
5
696
NM_007150.3
NP_009081.2
19
1792
XM_005274730.2
XP_005274787.1
21
1995
XM_005274731.2
XP_005274788.1
21
1992
XM_005274735.2
XP_005274792.1
20
1905
XM_005274737.2
XP_005274794.1
20
1899
XM_005274738.2
XP_005274795.1
20
1896
XM_005274740.2
XP_005274797.1
19
1809
XM_005274741.2
XP_005274798.1
19
1809
XM_005274742.2
XP_005274799.1
19
1806
XM_005274744.2
XP_005274801.1
18
1722
XM_005274745.2
XP_005274802.1
18
1719
XM_005274746.3
XP_005274803.1
17
1638
XM_011531194.1
XP_011529496.1
19
1800
XM_011531195.1
XP_011529497.1
20
1797
XM_017029821.1
XP_016885310.1
22
2127
XM_017029822.1
XP_016885311.1
23
2147
XM_017029823.1
XP_016885312.1
22
2124
XM_017029824.1
XP_016885313.1
21
2043
XM_017029825.1
XP_016885314.1
21
2040
XM_017029826.1
XP_016885315.1
21
2037
XM_017029827.1
XP_016885316.1
21
2034
XM_017029828.1
XP_016885317.1
21
2031
XM_017029829.1
XP_016885318.1
20
1950
XM_017029830.1
XP_016885319.1
20
1944
XM_017029831.1
XP_016885320.1
20
1941
XM_017029832.1
XP_016885321.1
20
1932
XM_017029833.1
XP_016885322.1
19
1866
XM_017029834.1
XP_016885323.1
19
1854
XM_017029835.1
XP_016885324.1
19
1815
GenBank mRNA
AK056517.1
-
8
851
AK095258.1
-
5
668
AK296967.1
-
19
1795
AK301530.1
-
11
1345
AK304323.1
-
15
1358
AY997296.1
-
19
1781
BX537525.1
-
5
2370
BX640940.1
-
11
1092
DQ508022.1
-
19
1744
DQ508023.1
-
17
1567
DQ508024.1
-
18
1657
DQ508025.1
-
20
1840
DQ508026.1
-
19
1753
Y09538.1
-
11
1073
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline