Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023560.3 | NP_001018854.2 | 4 | 1370 | ||
NM_001111039.2 | NP_001104509.1 | 4 | 1367 | |||
NM_001287421.1 | NP_001274350.1 | 4 | 856 | |||
NM_001287422.1 | NP_001274351.1 | 2 | 766 | |||
NM_152736.5 | NP_689949.3 | 4 | 965 | |||
XM_011514862.2 | XP_011513164.1 | 4 | 1569 | |||
XM_011514864.1 | XP_011513166.1 | 5 | 1146 | |||
XM_011514866.1 | XP_011513168.1 | 4 | 918 | |||
XM_011514867.1 | XP_011513169.1 | 4 | 1282 | |||
XM_011514869.1 | XP_011513171.1 | 5 | 1210 | |||
XM_017011264.1 | XP_016866753.1 | 4 | 1567 | |||
XM_017011265.1 | XP_016866754.1 | 4 | 1301 | |||
GenBank mRNA | AK095948.1 | - | 4 | 917 | ||
AK293524.1 | - | 4 | 1090 | |||
AK297613.1 | - | 2 | 627 | |||
AK315645.1 | - | 4 | 829 | |||
AL832741.1 | - | 3 | 1558 | |||
BC013951.2 | - | 4 | 821 | 2963683 | ||
BC013962.2 | - | 4 | 821 | 2963683 | ||
BX537535.1 | - | 4 | 1239 | |||
Z11773.1 | - | 3 | 887 |