Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYM2 SIRNA Assays ›
Gene Symbol:
ZMYM2
Gene Name:
zinc finger MYM-type containing 2
Gene Aliases:
FIM, MYM, RAMP, SCLL, ZNF198
Chromosome Location:
Chr.13: 19957412 - 20091845 on Build GRCh38
UniGene ID:
Hs.507433
Species:
Human
siRNA ID
HSS187844
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190964.2
NP_001177893.1
19
3179
NM_001190965.2
NP_001177894.1
18
3194
NM_003453.4
NP_003444.1
19
3287
NM_197968.3
NP_932072.1
18
3240
XM_005266520.3
XP_005266577.1
18
3199
XM_011535216.2
XP_011533518.1
19
2869
XM_011535222.1
XP_011533524.1
18
3130
XM_011535223.2
XP_011533525.1
12
5584
XM_011535224.2
XP_011533526.1
12
1485
XM_017020728.1
XP_016876217.1
18
3130
XM_017020729.1
XP_016876218.1
18
3105
XM_017020731.1
XP_016876220.1
19
2922
XM_017020732.1
XP_016876221.1
19
2944
XM_017020733.1
XP_016876222.1
20
2971
XM_017020734.1
XP_016876223.1
20
2957
XM_017020735.1
XP_016876224.1
19
2921
XM_017020736.1
XP_016876225.1
19
3093
XM_017020737.1
XP_016876226.1
19
2868
XM_017020738.1
XP_016876227.1
18
3183
GenBank mRNA
AF012126.1
-
12
1582
AF060181.1
-
16
2537
AJ224901.1
-
19
3173
AK226118.1
-
18
3174
AK289741.1
-
18
3165
BC036372.1
-
18
3240
4823008
BX647944.1
-
18
3194
BX648905.1
-
11
3624
Y13472.1
-
19
3185
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cytoskeleton organization
multicellular organism development
biological_process
peptidyl-tyrosine phosphorylation
regulation of cell morphogenesis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein tyrosine kinase activity
zinc ion binding
ubiquitin conjugating enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline