Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145446.1 | NP_001138918.1 | 5 | 1107 | ||
NM_001145447.1 | NP_001138919.1 | 5 | 1312 | |||
NM_001145448.1 | NP_001138920.1 | 5 | 1231 | |||
NM_003454.3 | NP_003445.2 | 5 | 1315 | |||
NM_198087.2 | NP_932353.1 | 5 | 1107 | |||
NM_198088.2 | NP_932354.1 | 5 | 1234 | |||
XM_005255556.1 | XP_005255613.1 | 6 | 1267 | |||
XM_006720940.2 | XP_006721003.1 | 5 | 1638 | |||
XM_006720941.2 | XP_006721004.1 | 5 | 1110 | |||
XM_017023647.1 | XP_016879136.1 | 6 | 1406 | |||
GenBank mRNA | AB209219.1 | - | 1 | 3405 | ||
AF060866.1 | - | 5 | 919 | |||
AK056010.1 | - | 5 | 1312 | |||
AK315036.1 | - | 5 | 845 | |||
BC012909.1 | - | 5 | 1296 | 4470685 | ||
BC032575.1 | - | 5 | 1233 | 5490702 | ||
BC054005.1 | - | 5 | 1061 | 6166814 | ||
Y14443.1 | - | 5 | 1010 |