Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193511.1 | NP_001180440.1 | 10 | 1824 | ||
NM_006301.3 | NP_006292.3 | 11 | 1725 | |||
XM_005269138.3 | XP_005269195.1 | 10 | 1752 | |||
XM_006719588.3 | XP_006719651.1 | 10 | 1646 | |||
XM_011538725.2 | XP_011537027.1 | 10 | 2629 | |||
XM_017019956.1 | XP_016875445.1 | 11 | 1653 | |||
GenBank mRNA | AB209453.1 | - | 4 | 2795 | ||
AK094195.1 | - | 10 | 1620 | |||
AK314530.1 | - | 10 | 1983 | |||
U07358.1 | - | 11 | 1552 |