Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNALI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003462.3 | NP_003453.2 | 5 | 802 | ||
XM_005271172.2 | XP_005271229.1 | 4 | 614 | |||
GenBank mRNA | AF006386.1 | - | 5 | 779 | ||
AI128097.1 | - | 2 | 170 | |||
AK126963.1 | - | 1 | 1693 | |||
AK225935.1 | - | 4 | 2841 | |||
AK290502.1 | - | 5 | 726 | |||
AK295199.1 | - | 4 | 545 | |||
BC039074.1 | - | 5 | 785 | 5179639 | ||
BC046117.1 | - | 5 | 752 | 6192652 | ||
BX647913.1 | - | 5 | 802 |