Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018054.2 | NP_001018064.1 | 17 | 2892 | ||
NM_004631.4 | NP_004622.2 | 17 | 2892 | |||
NM_017522.4 | NP_059992.3 | 15 | 2280 | |||
NM_033300.3 | NP_150643.2 | 15 | 2382 | |||
XM_005271173.3 | XP_005271230.1 | 18 | 2841 | |||
XM_005271174.3 | XP_005271231.1 | 17 | 2449 | |||
XM_005271175.3 | XP_005271232.1 | 16 | 2410 | |||
XM_006710881.3 | XP_006710944.1 | 18 | 2841 | |||
XM_006710882.3 | XP_006710945.1 | 17 | 2616 | |||
XM_011542094.2 | XP_011540396.1 | 17 | 2715 | |||
XM_011542095.2 | XP_011540397.1 | 16 | 2320 | |||
XM_017002265.1 | XP_016857754.1 | 16 | 2676 | |||
XM_017002266.1 | XP_016857755.1 | 17 | 2616 | |||
XM_017002267.1 | XP_016857756.1 | 16 | 2286 | |||
XM_017002268.1 | XP_016857757.1 | 17 | 2325 | |||
GenBank mRNA | AK096482.1 | - | 15 | 2160 | ||
BC006443.1 | - | 4 | 498 | |||
BC051836.1 | - | 17 | 2797 | 6143442 | ||
D50678.1 | - | 17 | 2757 | |||
Z75190.1 | - | 15 | 2040 |