Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DDR1 SIRNA Assays ›
Gene Symbol:
DDR1
Gene Name:
discoidin domain receptor tyrosine kinase 1
Gene Aliases:
CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
Chromosome Location:
Chr.6: 30880909 - 30900156 on Build GRCh38
UniGene ID:
Hs.631988
Species:
Human
siRNA ID
HSS187879
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DDR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001202521.1
NP_001189450.1
11
1641
NM_001202522.1
NP_001189451.1
11
1559
NM_001202523.1
NP_001189452.1
13
2138
NM_001297652.1
NP_001284581.1
13
2257
NM_001297653.1
NP_001284582.1
13
2059
NM_001297654.1
NP_001284583.1
14
2170
NM_001954.4
NP_001945.3
15
2177
NM_013993.2
NP_054699.2
15
2214
NM_013994.2
NP_054700.2
13
1997
XM_006715185.2
XP_006715248.1
14
2161
XM_011514882.2
XP_011513184.1
14
2161
XM_011514883.1
XP_011513185.1
16
2276
XM_011514884.1
XP_011513186.1
16
2352
XM_011514885.1
XP_011513187.1
14
2199
XM_011514886.1
XP_011513188.1
14
2064
XM_011514887.1
XP_011513189.1
14
2214
XM_011514888.2
XP_011513190.1
13
2050
XM_017011268.1
XP_016866757.1
14
2031
XM_017011269.1
XP_016866758.1
13
2103
GenBank mRNA
AB210021.1
-
14
1998
AK130776.1
-
9
1151
AK291621.1
-
13
1952
AK294793.1
-
13
2124
BC008716.2
-
15
2177
2964574
BC013400.2
-
15
2177
2964574
BC070070.1
-
13
1948
5262509
BI193181.1
-
1
40
L11315.1
-
15
2096
L20817.1
-
14
2147
L57508.1
-
13
2036
X74979.1
-
12
1889
Z29093.1
-
12
2180
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
regulation of cell-matrix adhesion
cell adhesion
embryo implantation
lactation
negative regulation of cell proliferation
regulation of extracellular matrix disassembly
smooth muscle cell migration
extracellular matrix organization
organ regeneration
collagen-activated tyrosine kinase receptor signaling pathway
peptidyl-tyrosine autophosphorylation
ear development
skin development
wound healing, spreading of cells
protein autophosphorylation
branching involved in mammary gland duct morphogenesis
mammary gland alveolus development
smooth muscle cell-matrix adhesion
transmembrane receptor protein tyrosine kinase activity
protein binding
collagen binding
ATP binding
protein tyrosine kinase collagen receptor activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-fls9w:80/100.66.78.88:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline