Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199849.1 | NP_001186778.1 | 8 | 742 | ||
NM_001199850.1 | NP_001186779.1 | 6 | 547 | |||
NM_001199851.1 | NP_001186780.1 | 6 | 526 | |||
NM_004632.3 | NP_004623.1 | 7 | 646 | |||
NM_033657.2 | NP_387506.1 | 7 | 715 | |||
XM_005245480.2 | XP_005245537.1 | 6 | 612 | |||
XM_005245481.2 | XP_005245538.1 | 6 | 591 | |||
XM_005245482.4 | XP_005245539.1 | 5 | 426 | |||
XM_011509972.2 | XP_011508274.1 | 7 | 645 | |||
XM_017002289.1 | XP_016857778.1 | 7 | 714 | |||
XM_017002290.1 | XP_016857779.1 | 8 | 738 | |||
XM_017002291.1 | XP_016857780.1 | 7 | 632 | |||
XM_017002292.1 | XP_016857781.1 | 7 | 611 | |||
XM_017002293.1 | XP_016857782.1 | 6 | 530 | |||
XM_017002294.1 | XP_016857783.1 | 7 | 632 | |||
XM_017002295.1 | XP_016857784.1 | 7 | 611 | |||
XM_017002296.1 | XP_016857785.1 | 6 | 591 | |||
XM_017002297.1 | XP_016857786.1 | 5 | 489 | |||
GenBank mRNA | AK293117.1 | - | 7 | 647 | ||
AK298201.1 | - | 6 | 534 | |||
AK302281.1 | - | 6 | 547 | |||
BC002358.1 | - | 6 | 674 | |||
BC107487.1 | - | 7 | 646 | 40032516 | ||
BC107488.1 | - | 7 | 715 | 40032512 | ||
BT019494.1 | - | 6 | 558 | |||
CR749790.1 | - | 7 | 726 | |||
U18321.1 | - | 7 | 727 | |||
X83544.1 | - | 7 | 631 |