Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015120.4 | NP_055935.4 | 13 | 10098 | ||
GenBank mRNA | AB002326.3 | - | 6 | 3787 | ||
AJ417593.2 | - | 13 | 10098 | |||
AL833495.1 | - | 6 | 2930 | |||
AX751581.1 | - | 12 | 9972 | |||
BC035025.2 | - | 6 | 2786 | |||
BC050330.1 | - | 6 | 2786 | |||
BX647706.1 | - | 6 | 4011 | |||
BX648632.1 | - | 6 | 4511 |