Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308217.1 | NP_001295146.1 | 4 | 462 | ||
NM_001308222.1 | NP_001295151.1 | 4 | 1215 | |||
NM_003471.3 | NP_003462.2 | 4 | 582 | |||
NM_172159.3 | NP_751891.1 | 4 | 1265 | |||
NM_172160.2 | NP_751892.1 | 4 | 462 | |||
XM_011513115.2 | XP_011511417.1 | 4 | 175 | |||
XM_011513116.2 | XP_011511418.1 | 2 | 254 | |||
XM_011513117.2 | XP_011511419.1 | 2 | 65 | |||
XM_017007171.1 | XP_016862660.1 | 4 | 252 | |||
XM_017007172.1 | XP_016862661.1 | 4 | 1468 | |||
XM_017007173.1 | XP_016862662.1 | 2 | 169 | |||
XM_017007174.1 | XP_016862663.1 | 4 | 448 | |||
GenBank mRNA | AK057059.1 | - | 4 | 998 | ||
AK127240.1 | - | 4 | 1195 | |||
AK292693.1 | - | 4 | 448 | |||
AK292999.1 | - | 4 | 514 | |||
AK296728.1 | - | 2 | 166 | |||
AK303287.1 | - | 4 | 432 | |||
AY780786.1 | - | 4 | 344 | |||
BC043166.2 | - | 4 | 1215 | 5286833 | ||
DC410397.1 | - | 4 | 462 | |||
L39833.1 | - | 4 | 453 | |||
L47665.1 | - | 4 | 451 | |||
U16953.1 | - | 4 | 432 | |||
U17968.1 | - | 4 | 357 | |||
U33428.1 | - | 4 | 371 |