Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002876.2 | NP_001002876.1 | 5 | 473 | ||
NM_001110215.2 | NP_001103685.1 | 2 | 156 | |||
NM_001304370.1 | NP_001291299.1 | 5 | 678 | |||
NM_001304372.1 | NP_001291301.1 | 5 | 763 | |||
NM_001304373.1 | NP_001291302.1 | 4 | 586 | |||
NM_024053.4 | NP_076958.1 | 6 | 565 | |||
GenBank mRNA | AK123479.1 | - | 6 | 1590 | ||
AL160131.1 | - | 6 | 513 | |||
BC000705.2 | - | 6 | 512 | 3349507 | ||
BC007495.2 | - | 6 | 525 | 2966541 | ||
BE789455.1 | - | 4 | 518 | |||
BE797487.1 | - | 5 | 427 | |||
BG476718.1 | - | 4 | 556 | |||
BQ652721.1 | - | 5 | 515 | |||
BQ922403.1 | - | 6 | 498 | |||
BU556693.1 | - | 6 | 565 | |||
BX332140.2 | - | 4 | 523 | |||
BX354019.2 | - | 5 | 611 | |||
BX417558.2 | - | 4 | 524 | |||
CR456399.1 | - | 6 | 538 | |||
CR457392.1 | - | 6 | 477 | |||
EU035297.1 | - | 2 | 156 |