Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009960.1 | NP_001009960.1 | 3 | 1324 | ||
NM_001083956.1 | NP_001077425.1 | 4 | 1602 | |||
NM_001085368.1 | NP_001078837.1 | 4 | 1429 | |||
NM_138494.2 | NP_612503.1 | 3 | 1497 | |||
XM_017012598.1 | XP_016868087.1 | 6 | 2502 | |||
XM_017012599.1 | XP_016868088.1 | 6 | 1660 | |||
XM_017012600.1 | XP_016868089.1 | 6 | 1673 | |||
XM_017012601.1 | XP_016868090.1 | 6 | 1659 | |||
XM_017012602.1 | XP_016868091.1 | 5 | 2397 | |||
XM_017012603.1 | XP_016868092.1 | 4 | 2522 | |||
XM_017012604.1 | XP_016868093.1 | 4 | 1439 | |||
XM_017012605.1 | XP_016868094.1 | 5 | 1450 | |||
XM_017012606.1 | XP_016868095.1 | 3 | 1355 | |||
XM_017012607.1 | XP_016868096.1 | 3 | 1341 | |||
XM_017012608.1 | XP_016868097.1 | 6 | 2535 | |||
XM_017012609.1 | XP_016868098.1 | 5 | 1549 | |||
XM_017012610.1 | XP_016868099.1 | 5 | 1496 | |||
GenBank mRNA | AK122929.1 | - | 1 | 581 | ||
AK128057.1 | - | 2 | 2561 | |||
AK304785.1 | - | 4 | 1306 | |||
AK314593.1 | - | 3 | 1340 | |||
AK316024.1 | - | 4 | 1420 | |||
AY099353.1 | - | 3 | 1323 | |||
BC011816.1 | - | 1 | 592 | |||
BC024770.1 | - | 1 | 843 | |||
BC037407.1 | - | 3 | 1287 | 5752559 | ||
CR749270.1 | - | 3 | 1105 |