Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM38A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024074.2 | NP_076979.1 | 5 | 717 | ||
GenBank mRNA | AK025981.1 | - | 5 | 717 | ||
AK292764.1 | - | 5 | 702 | |||
BC001195.1 | - | 5 | 633 | 3355479 |