Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024094.2 | NP_076999.2 | 7 | 1025 | ||
XM_005251065.3 | XP_005251122.1 | 6 | 881 | |||
GenBank mRNA | AK054585.1 | - | 7 | 997 | ||
AK074355.1 | - | 7 | 962 | |||
AK315356.1 | - | 7 | 951 | |||
AY358866.1 | - | 7 | 977 | |||
BC001316.1 | - | 7 | 1004 | 3454505 | ||
BC001531.2 | - | 7 | 963 | 3510131 |