Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300925.1 | NP_001287854.1 | 4 | 243 | ||
NM_024104.3 | NP_077009.2 | 4 | 243 | |||
GenBank mRNA | AK025602.1 | - | 4 | 200 | ||
AL702263.1 | - | 4 | 244 | |||
BC001680.2 | - | 1 | 15 | |||
BC001948.1 | - | 1 | 15 | |||
BC069279.1 | - | 4 | 216 | |||
DB047166.1 | - | 4 | 228 |