Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270891.1 | NP_001257820.1 | 2 | 121 | ||
NM_024108.2 | NP_077013.1 | 2 | 163 | |||
XM_017027295.1 | XP_016882784.1 | 2 | 222 | |||
GenBank mRNA | AF161407.1 | - | 2 | 115 | ||
BC001907.1 | - | 2 | 147 | 3534111 | ||
BC004450.1 | - | 2 | 148 | 2820830 | ||
CN256687.1 | - | 2 | 133 | |||
FJ418644.1 | - | 2 | 103 |