Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306079.1 | NP_001293008.1 | 21 | 3570 | ||
NM_004341.4 | NP_004332.2 | 22 | 3759 | |||
XM_005264555.3 | XP_005264612.1 | 22 | 3892 | |||
XM_006712101.2 | XP_006712164.1 | 21 | 3703 | |||
GenBank mRNA | AK226105.1 | - | 21 | 3552 | ||
BC065510.1 | - | 22 | 3759 | 5551082 | ||
BU541191.1 | - | 1 | 23 | |||
D78586.1 | - | 22 | 3623 |