Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161527.1 | NP_001154999.1 | 6 | 899 | ||
NM_001292016.1 | NP_001278945.1 | 5 | 936 | |||
NM_024309.3 | NP_077285.3 | 6 | 1185 | |||
GenBank mRNA | AF372839.1 | - | 6 | 1185 | ||
AJ304866.1 | - | 6 | 1132 | |||
AK026176.1 | - | 6 | 898 | |||
AK096296.1 | - | 6 | 1176 | |||
AL137262.1 | - | 1 | 1719 | |||
BC002740.2 | - | 6 | 1143 | 3632736 | ||
CB157550.1 | - | 2 | 150 |