Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126339.3 | NP_001119811.1 | 2 | 643 | ||
NM_001256062.2 | NP_001242991.1 | 1 | 1042 | |||
NM_001256321.2 | NP_001243250.1 | 2 | 643 | |||
NM_024328.5 | NP_077304.1 | 1 | 1042 | |||
GenBank mRNA | AF432862.1 | - | 1 | 311 | ||
AK057691.1 | - | 1 | 749 | |||
AK098604.1 | - | 2 | 432 | |||
BC002984.1 | - | 1 | 736 | 3535451 | ||
BI522644.1 | - | 2 | 447 | |||
BM925597.1 | - | 2 | 630 | |||
BX161435.1 | - | 1 | 1002 | |||
BX378775.2 | - | 1 | 730 | |||
DB122722.1 | - | 2 | 354 |