Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024333.2 | NP_077309.1 | 12, 13 | 1523 | ||
XM_005259644.2 | XP_005259701.1 | 12, 13 | 1431 | |||
GenBank mRNA | AF316829.1 | - | 12, 13 | 1414 | ||
AK021750.1 | - | 12, 13 | 1423 | |||
AK315661.1 | - | 12, 13 | 1517 | |||
AY032617.1 | - | 12, 13 | 1446 | |||
BC003124.1 | - | 12, 13 | 1423 | 3502614 | ||
BC016442.1 | - | 6, 7 | 1127 | |||
CA749067.1 | - | 1, 2 | 341 |