Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014234.4 | NP_055049.1 | 3 | 391 | ||
GenBank mRNA | BC008185.1 | - | 3 | 386 | 4213813 | |
BT007239.1 | - | 3 | 359 | |||
HY167181.1 | - | 3 | 391 |